Learn More
Abnova™ Human LOC644068 Full-length ORF (XP_935199.1, 1 a.a. - 170 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00644068-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene is one of the multiple ribosomal protein S14-like genes that are dispersed throughout the genome. This gene is intronless, and may possibly be a pseudogene and/or a null allele of the spliced and functional ribosomal protein S14 gene that is located on chromosome 5. This intronless gene is transcribed and has the potential to encode a protein similar to ribosomal protein S14, but it is unclear as to whether or not a protein is produced. [provided by RefSeq]
Sequence: MAPGKGKEKKEEQVINLGPQVAEGENVFGVCHIFASFNDTFVHVTDLSGKQTICRVTGGMKVKADRDESSPYAAMLTTQDVAQRCKELGIIALHIQLRATGGNRTKTLGPGAQSALRALACSGMKIGRIEDVTPIPSDSTLRKGVTVVAVCEQDSSKYFLLINCLHVKNKSpecifications
XP_935199.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
44.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC87895 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
644068 | |
LOC644068 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAPGKGKEKKEEQVINLGPQVAEGENVFGVCHIFASFNDTFVHVTDLSGKQTICRVTGGMKVKADRDESSPYAAMLTTQDVAQRCKELGIIALHIQLRATGGNRTKTLGPGAQSALRALACSGMKIGRIEDVTPIPSDSTLRKGVTVVAVCEQDSSKYFLLINCLHVKNK | |
RUO | |
MGC87895 | |
Recombinant | |
wheat germ expression system |