Learn More
Abnova™ Human LMCD1 Full-length ORF (NP_055398.1, 1 a.a. - 365 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00029995-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene contains a cysteine-rich domain in the N-terminal region and 2 LIM domains in the C-terminal region. It also has several potential phosphorylation and N-myristoylation sites and a single potential N-glycosylation site. The presence of LIM domains implies involvement in protein-protein interactions. Expression of this gene has been detected in most tissues, with highest expression in skeletal muscle. Transcript variants utilizing alternative polyA signals have been observed. [provided by RefSeq]
Sequence: MAKVAKDLNPGVKKMSLGQLQSARGVACLGCKGTCSGFEPHSWRKICKSCKCSQEDHCLTSDLEDDRKIGRLLMDSKYSTLTARVKGGDGIRIYKRNRMIMTNPIATGKDPTFDTITYEWAPPGVTQKLGLQYMELIPKEKQPVTGTEGAFYRRRQLMHQLPIYDQDPSRCRGLLENELKLMEEFVKQYKSEALGVGEVALPGQGGLPKEEGKQQEKPEGAETTAATTNGSLSDPSKEVEYVCELCKGAAPPDSPVVYSDRAGYNKQWHPTCFVCAKCSEPLVDLIYFWKDGAPWCGRHYCESLRPRCSGCDEIIFAEDYQRVEDLAWHRKHFVCEGCEQLLSGRAYIVTKGQLLCPTCSKSKRSSpecifications
NP_055398.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
67.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LMCD1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
29995 | |
LMCD1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAKVAKDLNPGVKKMSLGQLQSARGVACLGCKGTCSGFEPHSWRKICKSCKCSQEDHCLTSDLEDDRKIGRLLMDSKYSTLTARVKGGDGIRIYKRNRMIMTNPIATGKDPTFDTITYEWAPPGVTQKLGLQYMELIPKEKQPVTGTEGAFYRRRQLMHQLPIYDQDPSRCRGLLENELKLMEEFVKQYKSEALGVGEVALPGQGGLPKEEGKQQEKPEGAETTAATTNGSLSDPSKEVEYVCELCKGAAPPDSPVVYSDRAGYNKQWHPTCFVCAKCSEPLVDLIYFWKDGAPWCGRHYCESLRPRCSGCDEIIFAEDYQRVEDLAWHRKHFVCEGCEQLLSGRAYIVTKGQLLCPTCSKSKRS | |
RUO | |
LMCD1 | |
Recombinant | |
wheat germ expression system |