Learn More
Abnova™ Human LIME1 Full-length ORF (AAH17016.1, 1 a.a. - 295 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00054923-P01.25ug
Additional Details : Weight : 0.00010kg
Description
LIME1 is a raft-associated transmembrane adaptor phosphoprotein that is preferentially expressed in hemopoietic cells, particularly T cells (Brdickova et al., 2003 [PubMed 14610046]; Hur et al., 2003 [PubMed 14610044]).[supplied by OMIM]
Sequence: MGLPVSWAPPALWVLGCCALLLSLWALCTACRRPEDAVAPRKRARRQRARLQGSATAAEASLLRRTHLCSLSKSDTRLHELHRGPRSSRALRPASMDLLRPHWLEVSRDITGPQAAPSAFPHQELPRALPAAAATAGCAGLEATYSNVGLAALPGVSLAASPVVAEYARVQKRKGTHRSPQEPQQGKTEVTPAAQVDVLYSRVCKPKRRDPGPTTDPLDPKGQGAILALAGDLAYQTLPLRALDVDSGPLENVYESIRELGDPAGRSSTCGAGTPPASSCPSLGRGWRPLPASLPSpecifications
AAH17016.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
57.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ20406/LIME/LP8067/dJ583P15.4 | |
LIME1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
54923 | |
LIME1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGLPVSWAPPALWVLGCCALLLSLWALCTACRRPEDAVAPRKRARRQRARLQGSATAAEASLLRRTHLCSLSKSDTRLHELHRGPRSSRALRPASMDLLRPHWLEVSRDITGPQAAPSAFPHQELPRALPAAAATAGCAGLEATYSNVGLAALPGVSLAASPVVAEYARVQKRKGTHRSPQEPQQGKTEVTPAAQVDVLYSRVCKPKRRDPGPTTDPLDPKGQGAILALAGDLAYQTLPLRALDVDSGPLENVYESIRELGDPAGRSSTCGAGTPPASSCPSLGRGWRPLPASLP | |
RUO | |
LIME1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |