missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LIMD1 Partial ORF (NP_055055.1, 577 a.a. - 674 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008994-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16189705

  • 4115.00 NOK / 10µg
Estimated Shipment: 12-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: VDSENKIYCVRDYHKVLAPKCAACGLPILPPEGSDETIRVVSMDRDYHVECYHCEDCGLELNDEDGHRCYPLEDHLFCHSCHVKRLEKRPSSTALHQH
Specifications

Specifications

NP_055055.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VDSENKIYCVRDYHKVLAPKCAACGLPILPPEGSDETIRVVSMDRDYHVECYHCEDCGLELNDEDGHRCYPLEDHLFCHSCHVKRLEKRPSSTALHQH
RUO
LIMD1
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
8994
LIMD1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
LIMD1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers