Learn More
Abnova™ Human LILRA5 Full-length ORF (NP_067073.1, 1 a.a. - 299 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00353514-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the leukocyte immunoglobulin-like receptor (LIR) family. LIR family members are known to have activating and inibitory functions in leukocytes. Crosslink of this receptor protein on the surface of monocytes has been shown to induce calcium flux and secretion of several proinflammatory cytokines, which suggests the roles of this protein in triggering innate immune responses. This gene is one of the leukocyte receptor genes that form a gene cluster on the chromosomal region 19q13.4. Four alternatively spliced transcript variants encoding distinct isoforms have been described. [provided by RefSeq]
Sequence: MAPWSHPSAQLQPVGGDAVSPALMVLLCLGLSLGPRTHVQAGNLSKATLWAEPGSVISRGNSVTIRCQGTLEAQEYRLVKEGSPEPWDTQNPLEPKNKARFSIPSMTEHHAGRYRCYYYSPAGWSEPSDPLELVVTGFYNKPTLSALPSPVVTSGENVTLQCGSRLRFDRFILTEEGDHKLSWTLDSQLTPSGQFQALFPVGPVTPSHRWMLRCYGSRRHILQVWSEPSDLLEIPVSGAADNLSPSQNKSDSGTASHLQDYAVENLIRMGMAGLILVVLGILIFQDWHSQRSPQAAAGRSpecifications
NP_067073.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
59.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD85/CD85F/ILT11/LILRB7/LIR9 | |
LILRA5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
353514 | |
LILRA5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAPWSHPSAQLQPVGGDAVSPALMVLLCLGLSLGPRTHVQAGNLSKATLWAEPGSVISRGNSVTIRCQGTLEAQEYRLVKEGSPEPWDTQNPLEPKNKARFSIPSMTEHHAGRYRCYYYSPAGWSEPSDPLELVVTGFYNKPTLSALPSPVVTSGENVTLQCGSRLRFDRFILTEEGDHKLSWTLDSQLTPSGQFQALFPVGPVTPSHRWMLRCYGSRRHILQVWSEPSDLLEIPVSGAADNLSPSQNKSDSGTASHLQDYAVENLIRMGMAGLILVVLGILIFQDWHSQRSPQAAAGR | |
RUO | |
LILRA5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |