missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LILRA1/CD85i/LIR-6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
1885.00 NOK - 4810.00 NOK
Specifications
| Antigen | LIR-6/LILRA1 |
|---|---|
| Clone | 586439 |
| Dilution | Immunocytochemistry 8-25 μ/mL |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18730277
![]() |
R&D Systems
MAB30852-SP |
25 μg |
1885.00 NOK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18794487
![]() |
R&D Systems
MAB30852-100 |
100 μg |
4810.00 NOK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LIR-6/LILRA1 Monoclonal specifically detects LIR-6/LILRA1 in Human samples. It is validated for Immunocytochemistry.Specifications
| LIR-6/LILRA1 | |
| Immunocytochemistry 8-25 μ/mL | |
| Monoclonal | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CD85 antigen-like family member I, CD85i, CD85i antigen, leukocyte immunoglobulin-like receptor subfamily A member 1, leukocyte immunoglobulin-like receptor subfamily A member 1 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 1, LIR6, LIR-6, LIR-6Leukocyte immunoglobulin-like receptor 6, LIR6MGC126563 | |
| Mouse myeloma cell line, NS0-derived human LILRA1/CD85i/LIR-6, Pro17-Asn461, Accession No. O75019 | |
| Primary | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 586439 | |
| Immunocytochemistry | |
| Unconjugated | |
| Human | |
| O75019 | |
| 11024 | |
| IgG1 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human LILRA1/CD85i/LIR-6 in direct ELISAs. In direct ELISAs, no cross-reactivity with human LILRA2, LILRA3, LILRA4, LILRA5, LILRA6, LILRB2 and LILRB3 was observed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title