missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LGR6 Partial ORF (NP_067649, 403 a.a. - 496 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00059352-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16189376

  • 6245.00 NOK / 25µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the leucine-rich repeat-containing subgroup of the G protein-coupled 7-transmembrane protein superfamily. The encoded protein is a glycoprotein hormone receptor with a large N-terminal extracellular domain that contains leucine-rich repeats important for the formation of a horseshoe-shaped interaction motif for ligand binding. Alternative splicing of this gene results in multiple transcript variants. [provided by RefSeq]

Sequence: ALSQAFSKDSFPKLRILEVPYAYQCCPYGMCASFFKASGQWEAEDLHLDDEESSKRPLGLLARQAENHYDQDLDELQLEMEDSKPHPSVQCSPT
Specifications

Specifications

NP_067649
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.08kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
ALSQAFSKDSFPKLRILEVPYAYQCCPYGMCASFFKASGQWEAEDLHLDDEESSKRPLGLLARQAENHYDQDLDELQLEMEDSKPHPSVQCSPT
RUO
LGR6
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
59352
LGR6 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ14471/GPCR/VTS20631
LGR6
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers