missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LGR6 Full-length ORF (NP_001017404.1, 1 a.a. - 828 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00059352-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16193173

  • 6245.00 NOK / 25µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the leucine-rich repeat-containing subgroup of the G protein-coupled 7-transmembrane protein superfamily. The encoded protein is a glycoprotein hormone receptor with a large N-terminal extracellular domain that contains leucine-rich repeats important for the formation of a horseshoe-shaped interaction motif for ligand binding. Alternative splicing of this gene results in multiple transcript variants. [provided by RefSeq]

Sequence: MRLEGEGRSARAGQNLSRAGSARRGAPRDLSMNNLTELQPGLFHHLRFLEELRLSGNHLSHIPGQAFSGLYSLKILMLQNNQLGGIPAEALWELPSLQSLDLNYNKLQEFPVAIRTLGRLQELGFHNNNIKAIPEKAFMGNPLLQTIHFYDNPIQFVGRSAFQYLPKLHTLSLNGAMDIQEFPDLKGTTSLEILTLTRAGIRLLPSGMCQQLPRLRVLELSHNQIEELPSLHRCQKLEEIGLQHNRIWEIGADTFSQLSSLQALDLSWNAIRSIHPEAFSTLHSLVKLDLTDNQLTTLPLAGLGGLMHLKLKGNLALSQAFSKDSFPKLRILEVPYAYQCCPYGMCASFFKASGQWEAEDLHLDDEESSKRPLGLLARQAENHYDQDLDELQLEMEDSKPHPSVQCSPTPGPFKPCEYLFESWGIRLAVWAIVLLSVLCNGLVLLTVFAGGPVPLPPVKFVVGAIAGANTLTGISCGLLASVDALTFGQFSEYGARWETGLGCRATGFLAVLGSEASVLLLTLAAVQCSVSVSCVRAYGKSPSLGSVRAGVLGCLALAGLAAALPLASVGEYGASPLCLPYAPPEGQPAALGFTVALVMMNSFCFLVVAGAYIKLYCDLPRGDFEAVWDCAMVRHVAWLIFADGLLYCPVAFLSFASMLGLFPVTPEAVKSVLLVVLPLPACLNPLLYLLFNPHFRDDLRRLRPRAGDSGPLAYAAAGELEKSSCDSTQALVAFSDVDLILEASEAGRPPGLETYGFPSVTLISCQQPGAPRLEGSHCVEPEGNHFGNPQPSMDGELLLRAEGSTPAGGGLSGGGGFQPSGLAFASHV
Specifications

Specifications

NP_001017404.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
115.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ14471/GPCR/VTS20631
LGR6
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
59352
LGR6 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MRLEGEGRSARAGQNLSRAGSARRGAPRDLSMNNLTELQPGLFHHLRFLEELRLSGNHLSHIPGQAFSGLYSLKILMLQNNQLGGIPAEALWELPSLQSLDLNYNKLQEFPVAIRTLGRLQELGFHNNNIKAIPEKAFMGNPLLQTIHFYDNPIQFVGRSAFQYLPKLHTLSLNGAMDIQEFPDLKGTTSLEILTLTRAGIRLLPSGMCQQLPRLRVLELSHNQIEELPSLHRCQKLEEIGLQHNRIWEIGADTFSQLSSLQALDLSWNAIRSIHPEAFSTLHSLVKLDLTDNQLTTLPLAGLGGLMHLKLKGNLALSQAFSKDSFPKLRILEVPYAYQCCPYGMCASFFKASGQWEAEDLHLDDEESSKRPLGLLARQAENHYDQDLDELQLEMEDSKPHPSVQCSPTPGPFKPCEYLFESWGIRLAVWAIVLLSVLCNGLVLLTVFAGGPVPLPPVKFVVGAIAGANTLTGISCGLLASVDALTFGQFSEYGARWETGLGCRATGFLAVLGSEASVLLLTLAAVQCSVSVSCVRAYGKSPSLGSVRAGVLGCLALAGLAAALPLASVGEYGASPLCLPYAPPEGQPAALGFTVALVMMNSFCFLVVAGAYIKLYCDLPRGDFEAVWDCAMVRHVAWLIFADGLLYCPVAFLSFASMLGLFPVTPEAVKSVLLVVLPLPACLNPLLYLLFNPHFRDDLRRLRPRAGDSGPLAYAAAGELEKSSCDSTQALVAFSDVDLILEASEAGRPPGLETYGFPSVTLISCQQPGAPRLEGSHCVEPEGNHFGNPQPSMDGELLLRAEGSTPAGGGLSGGGGFQPSGLAFASHV
RUO
LGR6
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers