missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human LAG-1 (CCL4L1) Recombinant Protein, PeproTech®

Product Code. p-7246066
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
5µg
This item is not returnable. View return policy

Product Code. 17842483

Brand: Gibco™ 30058100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human LAG-1 is a 7.7 kDa protein containing 69 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

LAG-1 is CC chemokine that signals through the CCR5 receptor. LAG-1 is identical to MIP-1beta (ACT II isotype) except for two amino acid substitutions; arginine for histidine at position 22 and serine for glycine at position 47 of the mature protein. LAG-1 chemoattracts monocytes, and exhibits activity as an HIV-suppressive factor.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8NHW4
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 388372
Molecular Weight (g/mol) 7.7 kDa
Name Human LAG-1 (CCL4L1)
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Common Name CCL4L1
Gene Symbol CCL4L1
Biological Activity Determined by its ability to chemoattract CCR5-transfected cells using a concentration of 0.1-10.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence APMGSDPPTA CCFSYTARKL PRNFVVDYYE TSSLCSQPAV VFQTKRSKQV CADPSESWVQ EYVYDLELN
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.