Learn More
Abnova™ Human KRT71 Full-length ORF (AAI03919.1, 1 a.a. - 523 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00112802-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into epithelial keratins and hair keratins. This gene encodes a protein that is expressed in the inner root sheath of hair follicles. The type II keratins are clustered in a region of chromosome 12q13
Sequence: MSRQFTCKSGAAAKGGFSGCSAVLSGGSSSSFRAGSKGLSGGFGSRSLYSLGGVRSLNVASGSGKSGGYGFGRGRASGFAGSMFGSVALGPVCPTVCPPGGIHQVTVNESLLAPLNVELDPEIQKVRAQEREQIKALNNKFASFIDKVRFLEQQNQVLETKWELLQQLDLNNCKNNLEPILEGYISNLRKQLETLSGDRVRLDSELRNVQDVVEDYKKRYEEEINKRTAAENEFVLLKKDVDAAYANKVELQAKVESMDQEIKFFRCLFEAEITQIQSHISDMSVILSMDNNRNLDLDSIIDEVRTQYEEIALKSKAEAEALYQTKFQELQLAAGRHGDDLKNTKNEISELTRLIQRIRSEIENVKKQASNLETAIADAEQRGDNALKDARAKLDELEGALHQAKEELARMLREYQELMSLKLALDMEIATYRKLLESEECRMSGEFPSPVSISIISSTSGGSGYGFRPSMVSGGYVANSSNCISGVCSVRGGEGRSRGSANDYKDTLGKGSSLSAPSKKTSRSpecifications
AAI03919.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
83.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
K6IRS1/KRT6IRS/KRT6IRS1/MGC119390/MGC119391 | |
KRT71 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
112802 | |
KRT71 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSRQFTCKSGAAAKGGFSGCSAVLSGGSSSSFRAGSKGLSGGFGSRSLYSLGGVRSLNVASGSGKSGGYGFGRGRASGFAGSMFGSVALGPVCPTVCPPGGIHQVTVNESLLAPLNVELDPEIQKVRAQEREQIKALNNKFASFIDKVRFLEQQNQVLETKWELLQQLDLNNCKNNLEPILEGYISNLRKQLETLSGDRVRLDSELRNVQDVVEDYKKRYEEEINKRTAAENEFVLLKKDVDAAYANKVELQAKVESMDQEIKFFRCLFEAEITQIQSHISDMSVILSMDNNRNLDLDSIIDEVRTQYEEIALKSKAEAEALYQTKFQELQLAAGRHGDDLKNTKNEISELTRLIQRIRSEIENVKKQASNLETAIADAEQRGDNALKDARAKLDELEGALHQAKEELARMLREYQELMSLKLALDMEIATYRKLLESEECRMSGEFPSPVSISIISSTSGGSGYGFRPSMVSGGYVANSSNCISGVCSVRGGEGRSRGSANDYKDTLGKGSSLSAPSKKTSR | |
RUO | |
KRT71 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |