Learn More
Abnova™ Human KREMEN1 Partial ORF (NP_114434, 310 a.a. - 391 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00083999-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a high-affinity dickkopf homolog 1 (DKK1) transmembrane receptor that functionally cooperates with DKK1 to block wingless (WNT)/beta-catenin signaling. The encoded protein is a component of a membrane complex that modulates canonical WNT signaling through lipoprotein receptor-related protein 6 (LRP6). It contains extracellular kringle, WSC, and CUB domains. Alternatively spliced transcript variants encoding distinct isoforms have been observed for this gene. [provided by RefSeq]
Sequence: INQAQGFAVLYQAVKEELPQERPAVNQTVAEVITEQANLSVSAARSSKVLYVITTSPSHPPQTVPGSNSWAPPMGAGSHRVESpecifications
NP_114434 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.76kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
INQAQGFAVLYQAVKEELPQERPAVNQTVAEVITEQANLSVSAARSSKVLYVITTSPSHPPQTVPGSNSWAPPMGAGSHRVE | |
RUO | |
KREMEN1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
83999 | |
KREMEN1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ31863/KREMEM1/KREMEN/KRM1 | |
KREMEN1 | |
Recombinant | |
wheat germ expression system |