missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KLRC3 Partial ORF (NP_002252, 132 a.a. - 240 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003823-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16137161

  • 4115.00 NOK / 10µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Natural killer (NK) cells are lymphocytes that can mediate lysis of certain tumor cells and virus-infected cells without previous activation. They can also regulate specific humoral and cell-mediated immunity. NK cells preferentially express several calcium-dependent (C-type) lectins, which have been implicated in the regulation of NK cell function. KLRC3 is a member of the NKG2 group which are expressed primarily in natural killer (NK) cells and encodes a family of transmembrane proteins characterized by a type II membrane orientation (extracellular C terminus) and the presence of a C-type lectin domain. The NKG2 gene family is located within the NK complex, a region that contains several C-type lectin genes preferentially expressed on NK cells. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]

Sequence: GKERRTWEESLQACASKNSSSLLSIDNEEEMKFLASILPSSWIGVFRNSSHHPWVTINGLAFKHEIKDSDHAERNCAMLHVRGLISDQCGSSRIIRRGFIMLTRLVLNS
Specifications

Specifications

NP_002252
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GKERRTWEESLQACASKNSSSLLSIDNEEEMKFLASILPSSWIGVFRNSSHHPWVTINGLAFKHEIKDSDHAERNCAMLHVRGLISDQCGSSRIIRRGFIMLTRLVLNS
RUO
KLRC3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
3823
KLRC3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NKG2-E/NKG2E
KLRC3
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers