Learn More
Abnova™ Human KLK10 Partial ORF (NP_002767, 167 a.a. - 276 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_002767 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 5655 |
Molecular Weight (g/mol) | 37.84kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16193555
|
Abnova™
H00005655-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16103565
|
Abnova™
H00005655-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. Its encoded protein is secreted and may play a role in suppression of tumorigenesis in breast and prostate cancers. Alternate splicing of this gene results in multiple transcript variants encoding the same protein. [provided by RefSeq]
Sequence: DQCQVAGWGTTAARRVKYNKGLTCSSITILSPKECEVFYPGVVTNNMICAGLDRGQDPCQSDSGGPLVCDETLQGILSWGVYPCGSAQHPAVYTQICKYMSWINKVIRSNSpecifications
NP_002767 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NES1/PRSSL1 | |
KLK10 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
5655 | |
KLK10 (Human) Recombinant Protein (Q01) | |
DQCQVAGWGTTAARRVKYNKGLTCSSITILSPKECEVFYPGVVTNNMICAGLDRGQDPCQSDSGGPLVCDETLQGILSWGVYPCGSAQHPAVYTQICKYMSWINKVIRSN | |
RUO | |
KLK10 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |