missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KLHL23 Full-length ORF (NP_653312.2, 1 a.a. - 558 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00151230-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16188653

  • 6245.00 NOK / 25µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MALKGQEDYIYLFKDSTHPVDFLDAFRTFYLDGLFTDITLQCPSGIIFHCHRAVLAACSNYFKAMFTADMKEKFKNKIKLSGIHHDILEGLVNYAYTSQIEITKRNVQSLLEAADLLQFLSVKKACERFLVRHLDIDNCIGMHSFAEFHVCPELEKESRRILCSKFKEVWQQEEFLEISLEKFLFILSRKNLSVWKEEAIIEPVIKWTAHDVENRIECLYNLLSYINIDIDPVYLKTALGLQRSCLLTENKIRSLIYNALNPMHKEISQRSTATMYIIGGYYWHPLSEVHIWDPLTNVWIQGAEIPDYTRESYGVTCLGPNIYVTGGYRTDNIEALDTVWIYNSESDEWTEGLPMLNARYYHCAVTLGGCVYALGGYRKGAPAEEAEFYDPLKEKWIPIANMIKGVGNATACVLHDVIYVIGGHCGYRGSCTYDKVQSYNSDINEWSLITSSPHPEYGLCSVPFENKLYLVGGQTTITECYDPEQNEWREIAPMMERRMECGAVIMNGCIYVTGGYSYSKGTYLQSIEKYDPDLNKWEIVGNLPSAMRSHGCVCVYNV
Specifications

Specifications

NP_653312.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
90.3kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DITHP/FLJ37812/MGC22679/MGC2610
KLHL23
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
151230
KLHL23 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MALKGQEDYIYLFKDSTHPVDFLDAFRTFYLDGLFTDITLQCPSGIIFHCHRAVLAACSNYFKAMFTADMKEKFKNKIKLSGIHHDILEGLVNYAYTSQIEITKRNVQSLLEAADLLQFLSVKKACERFLVRHLDIDNCIGMHSFAEFHVCPELEKESRRILCSKFKEVWQQEEFLEISLEKFLFILSRKNLSVWKEEAIIEPVIKWTAHDVENRIECLYNLLSYINIDIDPVYLKTALGLQRSCLLTENKIRSLIYNALNPMHKEISQRSTATMYIIGGYYWHPLSEVHIWDPLTNVWIQGAEIPDYTRESYGVTCLGPNIYVTGGYRTDNIEALDTVWIYNSESDEWTEGLPMLNARYYHCAVTLGGCVYALGGYRKGAPAEEAEFYDPLKEKWIPIANMIKGVGNATACVLHDVIYVIGGHCGYRGSCTYDKVQSYNSDINEWSLITSSPHPEYGLCSVPFENKLYLVGGQTTITECYDPEQNEWREIAPMMERRMECGAVIMNGCIYVTGGYSYSKGTYLQSIEKYDPDLNKWEIVGNLPSAMRSHGCVCVYNV
RUO
KLHL23
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers