Learn More
Abnova™ Human KLF8 Full-length ORF (NP_009181.2, 1 a.a. - 359 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00011279-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein which is a member of the Sp/KLF family of transcription factors. Members of this family contain a C-terminal DNA-binding domain with three Kruppel-like zinc fingers. The encoded protein is thought to play an important role in the regulation of epithelial to mesenchymal transition, a process which occurs normally during development but also during metastasis. A pseudogene has been identified on chromosome 16. Alternative splicing results in multiple transcript variants. [provided by RefSeq]
Sequence: MVDMDKLINNLEVQLNSEGGSMQVFKQVTASVRNRDPPEIEYRSNMTSPTLLDANPMENPALFNDIKIEPPEELLASDFSLPQVEPVDLSFHKPKAPLQPASMLQAPIRPPKPQSSPQTLVVSTSTSDMSTSANIPTVLTPGSVLTSSQSTGSQQILHVIHTIPSVSLPNKMGGLKTIPVVVQSLPMVYTTLPADGGPAAITVPLIGGDGKNAGSVKVDPTSMSPLEIPSDSEESTIESGSSALQSLQGLQQEPAAMAQMQGEESLDLKRRRIHQCDFAGCSKVYTKSSHLKAHRRIHTGEKPYKCTWDGCSWKFARSDELTRHFRKHTGIKPFRCTDCNRSFSRSDHLSLHRRRHDTMSpecifications
NP_009181.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
65.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BKLF3/DKFZp686O08126/DXS741/MGC138314/ZNF741 | |
KLF8 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
11279 | |
KLF8 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MVDMDKLINNLEVQLNSEGGSMQVFKQVTASVRNRDPPEIEYRSNMTSPTLLDANPMENPALFNDIKIEPPEELLASDFSLPQVEPVDLSFHKPKAPLQPASMLQAPIRPPKPQSSPQTLVVSTSTSDMSTSANIPTVLTPGSVLTSSQSTGSQQILHVIHTIPSVSLPNKMGGLKTIPVVVQSLPMVYTTLPADGGPAAITVPLIGGDGKNAGSVKVDPTSMSPLEIPSDSEESTIESGSSALQSLQGLQQEPAAMAQMQGEESLDLKRRRIHQCDFAGCSKVYTKSSHLKAHRRIHTGEKPYKCTWDGCSWKFARSDELTRHFRKHTGIKPFRCTDCNRSFSRSDHLSLHRRRHDTM | |
RUO | |
KLF8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |