Learn More
Abnova™ Human KLC2 Full-length ORF (NP_073733.1, 1 a.a. - 622 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00064837-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Kinesin is a molecular motor that generates ATP-dependent movement of vesicles and organelles along microtubules. Kinesin consists of 2 light chains, such as KLC2, and 2 heavy chains (see KIF5B; MIM 602809) in a 1:1 stoichiometric ratio (Rahman et al., 1998 [PubMed 9624122]).[supplied by OMIM]
Sequence: MAMMVFPREEKLSQDEIVLGTKAVIQGLETLRGEHRALLAPLVAPEAGEAEPGSQERCILLRRSLEAIELGLGEAQVILALSSHLGAVESEKQKLRAQVRRLVQENQWLREELAGTQQKLQRSEQAVAQLEEEKQHLLFMSQIRKLDEDASPNEEKGDVPKDTLDDLFPNEDEQSPAPSPGGGDVSGQHGGYEIPARLRTLHNLVIQYASQGRYEVAVPLCKQALEDLEKTSGHDHPDVATMLNILALVYRDQNKYKEAAHLLNDALAIREKTLGKDHPAVAATLNNLAVLYGKRGKYKEAEPLCKRALEIREKVLGKFHPDVAKQLSNLALLCQNQGKAEEVEYYYRRALEIYATRLGPDDPNVAKTKNNLASCYLKQGKYQDAETLYKEILTRAHEKEFGSVNGDNKPIWMHAEEREESKDKRRDSAPYGEYGSWYKACKVDSPTVNTTLRSLGALYRRQGKLEAAHTLEDCASRNRKQGLDPASQTKVVELLKDGSGRRGDRRSSRDMAGGAGPRSESDLEDVGPTAEWNGDGSGSLRRSGSFGKLRDALRRSSEMLVKKLQGGTPQEPPNPRMKRASSLNFLNKSVEEPTQPGGTGLSDSRTLSSSSMDLSRRSSLVGSpecifications
NP_073733.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
95.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ12387 | |
KLC2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
64837 | |
KLC2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAMMVFPREEKLSQDEIVLGTKAVIQGLETLRGEHRALLAPLVAPEAGEAEPGSQERCILLRRSLEAIELGLGEAQVILALSSHLGAVESEKQKLRAQVRRLVQENQWLREELAGTQQKLQRSEQAVAQLEEEKQHLLFMSQIRKLDEDASPNEEKGDVPKDTLDDLFPNEDEQSPAPSPGGGDVSGQHGGYEIPARLRTLHNLVIQYASQGRYEVAVPLCKQALEDLEKTSGHDHPDVATMLNILALVYRDQNKYKEAAHLLNDALAIREKTLGKDHPAVAATLNNLAVLYGKRGKYKEAEPLCKRALEIREKVLGKFHPDVAKQLSNLALLCQNQGKAEEVEYYYRRALEIYATRLGPDDPNVAKTKNNLASCYLKQGKYQDAETLYKEILTRAHEKEFGSVNGDNKPIWMHAEEREESKDKRRDSAPYGEYGSWYKACKVDSPTVNTTLRSLGALYRRQGKLEAAHTLEDCASRNRKQGLDPASQTKVVELLKDGSGRRGDRRSSRDMAGGAGPRSESDLEDVGPTAEWNGDGSGSLRRSGSFGKLRDALRRSSEMLVKKLQGGTPQEPPNPRMKRASSLNFLNKSVEEPTQPGGTGLSDSRTLSSSSMDLSRRSSLVG | |
RUO | |
KLC2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |