Learn More
Abnova™ Human KIF3B Partial ORF (NP_004789.1, 648 a.a. - 747 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009371-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene acts as a heterodimer with kinesin family member 3A to aid in chromosome movement during mitosis and meiosis. The encoded protein is a plus end-directed microtubule motor and can interact with the SMC3 subunit of the cohesin complex. In addition, the encoded protein may be involved in the intracellular movement of membranous organelles. This protein and kinesin family member 3A form the kinesin II subfamily of the kinesin superfamily. [provided by RefSeq]
Sequence: MIRPEARYRAENIVLLELDMPSRTTRDYEGPAIAPKVQAALDAALQDEDEIQVDASSFESTANKKSKARPKSGRKSGSSSSSSGTPASQLYPQSRGLVPKSpecifications
NP_004789.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MIRPEARYRAENIVLLELDMPSRTTRDYEGPAIAPKVQAALDAALQDEDEIQVDASSFESTANKKSKARPKSGRKSGSSSSSSGTPASQLYPQSRGLVPK | |
RUO | |
KIF3B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9371 | |
KIF3B (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HH0048/KIAA0359 | |
KIF3B | |
Recombinant | |
wheat germ expression system |