Learn More
Abnova™ Human KIF2C Partial ORF (AAH14924, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00011004-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of kinesin-like protein family. Proteins of this family are microtubule-dependent molecular motors that transport organelles within cells and move chromosomes during cell division. This protein is important for anaphase chromosome segregation and may be required to coordinate the onset of sister centromere separation. [provided by RefSeq]
Sequence: MAMDSSLQARLFPGLAIKIQRSNGLIHSANVRTVNLEKSCVSVEWAEGGATKGKEIDFDDVAAINPELLQLLPLHPKDNLPLQENVTIQKQKRRSVNSKISpecifications
AAH14924 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAMDSSLQARLFPGLAIKIQRSNGLIHSANVRTVNLEKSCVSVEWAEGGATKGKEIDFDDVAAINPELLQLLPLHPKDNLPLQENVTIQKQKRRSVNSKI | |
RUO | |
KIF2C | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
11004 | |
KIF2C (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KNSL6/MCAK | |
KIF2C | |
Recombinant | |
wheat germ expression system |