Learn More
Abnova™ Human KIF21B Partial ORF (XP_371332, 1183 a.a. - 1282 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00023046-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
KIF21B belongs to a family of plus end-directed kinesin (see MIM 600025) motor proteins. Neurons use kinesin and dynein (see MIM 600112) microtubule-dependent motor proteins to transport essential cellular components along axonal and dendritic microtubules.[supplied by OMIM]
Sequence: VSRTVSLPTRGSTFPRQSRATETSPLTRRKSYDRGQPIRSTDVGFTPPSSPPTRPRNDRNVFSRLTSNQSQGSALDKSDDSDSSLSEVLRGIISPVGGAKSpecifications
XP_371332 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VSRTVSLPTRGSTFPRQSRATETSPLTRRKSYDRGQPIRSTDVGFTPPSSPPTRPRNDRNVFSRLTSNQSQGSALDKSDDSDSSLSEVLRGIISPVGGAK | |
RUO | |
KIF21B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
23046 | |
KIF21B (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ16314 | |
KIF21B | |
Recombinant | |
wheat germ expression system |