missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human KIF13B (aa 1737-1826) Control Fragment Recombinant Protein

Product Code. 30201733
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. Quantity unitSize
30201733 100 μL 100µL
1 options
This item is not returnable. View return policy

Product Code. 30201733

Brand: Invitrogen™ RP108203

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (87%), Rat (87%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83973 (PA5-83973. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

KIF13B (Kinesin Family Member 13B) is a Protein Coding gene. Diseases associated with KIF13B includeSpastic Paraplegia 37, Autosomal DominantandSpastic Paraplegia 11, Autosomal Recessive. Among its related pathways areGolgi-to-ER retrograde transportandFactors involved in megakaryocyte development and platelet production. Gene Ontology (GO) annotations related to this gene includeprotein kinase bindingandmicrotubule motor activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NQT8
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 23303
Name Human KIF13B (aa 1737-1826) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 5330429L19Rik; 6030414C01; AI429803; C130021D12Rik; GAKIN; guanylate kinase associated kinesin; KIAA0639; Kif13b; kinesin 13 B; kinesin family member 13 B; kinesin-like protein GAKIN; Kinesin-like protein KIF13B; N-3 kinesin
Common Name KIF13B
Gene Symbol KIF13B
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LPSGKNDGSIGGKQYFRCNPGYGLLVRPSRVRRATGPVRRRSTGLRLGAPEARRSATLSGSATNLASLTAALAKADRSHKNPENRKSWAS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.