Learn More
Abnova™ Human KIAA1958 Full-length ORF (NP_597722.1, 1 a.a. - 716 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00158405-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MEDCLHTSSENLSKLVSWAHSHGTICSLIPNLKHLLSEGSHGNLTAMWGCSAGHAYHWPLTATCRAGSQERVCFQDNRSFNSDSPSIIGVPSETQTSPVERYPGRPVKAKLDCNRTRDSCDFSYCSEPSELDETVEEYEDENTLFDMVCESSVTDEDSDFEPQTQRPQSIARKRPGVVPSSLHSSSQTQMVDECSNDVIIKKIKQEIPEDYYIVANAELTGGVDGPALSLTQMAKPKPQTHAGPSCVGSAKLIPHVTSAISTELDPHGMSASPSVISRPIVQKTARVSLASPNRGPPGTHGTNQQVAMQMPVSTSHPNKQISIPLSALQLPGQDEQVASEEFLSHLPSQVSSCEVALSPSVNTEPEVSSSQQQPPVAPAITTEATAQCIPAYSTKLNKFPVFNINDDLNDLCTSAVSPNTTKATRYALNVWRYWCMTNGLKDHTDITKIPAVKLNELLENFYVTVKKSDGSDFLATSLHAIRRGLDRILKNAGVGFSITSSTFSSSTKKLKEKLWVLSKAGMSGARSRNIVYFSLSDEEEMWQAGCLGDDSPITLLSTVVKYNSQYLNMRTLQEHADLMYGDIELLKDPQNQPYFARTDSVKRESRSGSTRVCHGKIYHEHSRGHKQCPYCLLYKYMYIHRPPTQMEAKSPFYLTARKEATDMGSVWYEEQRMGLRSLRGIVPNLAKKVKLENCENFTFVSFTQVSRRLGSHSCCQSpecifications
NP_597722.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
105.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ39294/MGC142075 | |
KIAA1958 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
158405 | |
KIAA1958 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEDCLHTSSENLSKLVSWAHSHGTICSLIPNLKHLLSEGSHGNLTAMWGCSAGHAYHWPLTATCRAGSQERVCFQDNRSFNSDSPSIIGVPSETQTSPVERYPGRPVKAKLDCNRTRDSCDFSYCSEPSELDETVEEYEDENTLFDMVCESSVTDEDSDFEPQTQRPQSIARKRPGVVPSSLHSSSQTQMVDECSNDVIIKKIKQEIPEDYYIVANAELTGGVDGPALSLTQMAKPKPQTHAGPSCVGSAKLIPHVTSAISTELDPHGMSASPSVISRPIVQKTARVSLASPNRGPPGTHGTNQQVAMQMPVSTSHPNKQISIPLSALQLPGQDEQVASEEFLSHLPSQVSSCEVALSPSVNTEPEVSSSQQQPPVAPAITTEATAQCIPAYSTKLNKFPVFNINDDLNDLCTSAVSPNTTKATRYALNVWRYWCMTNGLKDHTDITKIPAVKLNELLENFYVTVKKSDGSDFLATSLHAIRRGLDRILKNAGVGFSITSSTFSSSTKKLKEKLWVLSKAGMSGARSRNIVYFSLSDEEEMWQAGCLGDDSPITLLSTVVKYNSQYLNMRTLQEHADLMYGDIELLKDPQNQPYFARTDSVKRESRSGSTRVCHGKIYHEHSRGHKQCPYCLLYKYMYIHRPPTQMEAKSPFYLTARKEATDMGSVWYEEQRMGLRSLRGIVPNLAKKVKLENCENFTFVSFTQVSRRLGSHSCCQ | |
RUO | |
KIAA1958 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |