missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KIAA1201 Partial ORF (XP_370660, 250 a.a. - 353 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00057476-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16122713

  • 4115.00 NOK / 10µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: NTMGYCEEIPVEENEVNDSSSKSSIETKPDASPQLPKKSITNSTLTSTGSSEAPVSFDGLPLEEEALEGDGSLEKELAIDNIMGEKIEMIAPVNSPSLDFNDNE
Specifications

Specifications

XP_370660
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.18kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
NTMGYCEEIPVEENEVNDSSSKSSIETKPDASPQLPKKSITNSTLTSTGSSEAPVSFDGLPLEEEALEGDGSLEKELAIDNIMGEKIEMIAPVNSPSLDFNDNE
RUO
GRAMD1B
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
57476
KIAA1201 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA1201/MGC125622/MGC125623/MGC125625
GRAMD1B
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers