Learn More
Abnova™ Human JRKL Partial ORF (NP_003763, 426 a.a. - 524 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008690-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The function of this gene has not yet been defined, however, the encoded protein shares similarity with the human (41% identical) and mouse (34% identical) jerky gene products. This protein may act as a nuclear regulatory protein. [provided by RefSeq]
Sequence: ENLEKWLEVDSTEPGYEVLTDSEIIRRAQGQADESSENEEEEIELIPEKHINHAAALQWTENLLDYLEQQGDMILPDRLVIRKLRATIRNKQKMTKSSQSpecifications
NP_003763 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ENLEKWLEVDSTEPGYEVLTDSEIIRRAQGQADESSENEEEEIELIPEKHINHAAALQWTENLLDYLEQQGDMILPDRLVIRKLRATIRNKQKMTKSSQ | |
RUO | |
JRKL | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8690 | |
JRKL (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HHMJG/MGC129608 | |
JRKL | |
Recombinant | |
wheat germ expression system |