missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ITGA1 Partial ORF (ENSP00000282588, 951 a.a. - 1059 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003672-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16126991

  • 3655.00 NOK / 10µg
Estimated Shipment: 16-05-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes the alpha 1 subunit of integrin receptors. This protein heterodimerizes with the beta 1 subunit to form a cell-surface receptor for collagen and laminin. The heterodimeric receptor is involved in cell-cell adhesion and may play a role in inflammation and fibrosis. The alpha 1 subunit contains an inserted (I) von Willebrand factor type I domain which is thought to be involved in collagen binding. [provided by RefSeq]

Sequence: QFYSSASEYHISIAANETVPEVINSTEDIGNEINIFYLIRKSGSFPMPELKLSISFPNMTSNGYPVLYPTGLSSSENANCRPHIFEDPFSINSGKKMTTSTDHLKRGTI
Specifications

Specifications

ENSP00000282588
Liquid
3672
ITGA1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD49a/VLA1
ITGA1
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QFYSSASEYHISIAANETVPEVINSTEDIGNEINIFYLIRKSGSFPMPELKLSISFPNMTSNGYPVLYPTGLSSSENANCRPHIFEDPFSINSGKKMTTSTDHLKRGTI
RUO
ITGA1
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers