missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human IL22 Partial ORF (NP_065386, 80 a.a. - 179 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00050616-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: FHGVSMSERCYLMKQVLNFTLEEVLFPQSDRFQPYMQEVVPFLARLSNRLSTCHIEGDDLHIQRNVQKLKDTVKKLGESGEIKAIGELDLLFMSLRNACISpecifications
NP_065386 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
FHGVSMSERCYLMKQVLNFTLEEVLFPQSDRFQPYMQEVVPFLARLSNRLSTCHIEGDDLHIQRNVQKLKDTVKKLGESGEIKAIGELDLLFMSLRNACI | |
RUO | |
IL22 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
50616 | |
IL22 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IL-21/IL-22/IL-D110/IL-TIF/IL21/ILTIF/MGC79382/MGC79384/TIFIL-23/TIFa/zcyto18 | |
IL22 | |
Recombinant | |
wheat germ expression system |