missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-8 (CXCL8) (Monocyte-derived) Recombinant Protein

Recombinant IL8 Monocyte is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PHC0881

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 10477063

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Human IL-8 (CXCL8) (Monocyte-derived) Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

IL8 is chemotactic for all known types of migratory immune cells. IL8 also inhibits the adhesion of leukocytes to activated endothelial cells.

Shipping Condition: Wet Ice.

Specifications

Specifications

Cell Culture
8.6 kDa
100μg
Human
Carrier-Free
100 μg
E. coli
Recombinant, Ligand
Interleukins
Gibco™
3576
Sequential Chromatography
IL-8 Monocyte
Store in refrigerator (2–8°C).
< 0.1 ng/μg
Wet Ice
Chemokines & Receptors
Lyophilized
Immunology, Angiogenesis, Respiratory Research, Virology, Stem Cell Research, Inflammation
95 %
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.