missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL-31, AOF Recombinant Protein

E. coli Recombinant Protein

Brand:  Invitrogen™ RP87713

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 17616535

  • 11420.00 NOK / 100µg
Estimated Shipment: 04-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Interleukin-31 (IL-31) is a recently discovered T-cell cytokine closely related to IL-6 type cytokines and is preferentially produced by T helper type 2 cells. IL-31 activity is mediated through the ligand-induced oligomerization of a dimeric receptor complex containing IL-31 receptor A and oncostatin M receptor. In response to IL-31 binding, these proteins activate the JAK/STAT and the AKT signaling pathways. RNA levels of IL-31 receptor A and oncostatin M receptor are induced in activated monocytes but are expressed constitutively in epithelial cells. IL-31, when overexpressed in transgenic mice, results in the development of pruritis, alopecia, and skin lesions and in humans may result in atopic dermatitis, suggesting that IL-31 may represent a novel target for antipruritic drug development.
  • Purity: >95% by SDS-Page
  • Endotoxin Concentration: AX41 ≤EUs/μg
  • Protein Length: 141 aa.
  • Biological Activity: <25 ng/mL
  • Molecular Weight: 15.8 kDa
  • Animal Origin Free: Yes
Specifications

Specifications

Q6EBC2
20 mM sodium phosphate with 50 mM NaCl and no preservative
15.8 kDa
100 μg
-20°C
1700013B14Rik; Il31; IL-31; ILN; Interleukin; interleukin 31; Interleukin-31
Human
Recombinant
>95%
Control, ELISA, Functional Assay, Western Blot
386653
Human IL-31, AOF
E. coli
RUO
Il31
Unconjugated
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers