Learn More
ACROBiosystems Human IL-22 Protein, His Tag
Brand: ACROBiosystems IL2-H524a-1mg
Additional Details : Weight : 0.00010kg
Description
Interleukin-22 (IL22) is also known as cytokine Zcyto18, IL-10-related T-cell-derived-inducible factor (IL-TIF), which is belongs to the IL-10 family or IL-10 superfamily (including IL-19, IL-20, IL-24, and IL-26), a class of potent mediators of cellular inflammatory responses. IL-22 is produced by activated DC and T cells and initiates innate immune responses against bacterial pathogens especially in epithelial cells such as respiratory and gut epithelial cells. IL-22 biological activity is initiated by binding to a cell-surface complex composed of IL-22R1 and IL-10R2 receptor chains and further regulated by interactions with a soluble binding protein IL-22BP. IL-22 also promotes hepatocyte survival in the liver and epithelial cells in the lung and gut similar to IL-10.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 18.6 kDa. The protein migrates as 28-34 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_065386.1 | |
1 mg | |
Ala 34 - Ile 179 | |
IL-22,IL-TIF,ZCYTO18 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
18.6 kDa | |
Human IL-22, His Tag (IL2-H524a) is expressed from human 293 cells (HEK293). It contains AA Ala 34 - Ile 179 (Accession # Q9GZ x 6-1). | |
-20°C | |
IL-22 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Cytokines and Growth Factors |