missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-21R Antibody, R&D Systems™

Description
IL-21R Monoclonal specifically detects IL-21R in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
Specifications
| Antigen | IL-21R |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Clone | 152512 |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | Q9HBE5 |
| Gene Alias | CD360, CD360 antigen, IL-21 R, IL-21 receptor, IL21R, IL-21R, interleukin 21 receptor, interleukin-21 receptor, MGC10967, NILR, Novel interleukin receptor |
| Gene Symbols | IL21R |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?