missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-20 Recombinant Protein, PeproTech®

Product Code. 17830073
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
2µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 17830073

Brand: Gibco™ 20020500UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human IL-20 is a 35.2 kDa homodimeric protein consisting of two 153 amino acid polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Cytokines that signal through Class II receptors form a distinct family that includes the interferons and interleukin 10 (IL-10). Recent identification of several IL-10 homologs has defined a cytokine subfamily that includes AK155, IL-19, IL-20, IL-22, and IL-24. IL-20 belongs to the IL-10 family and plays a role in skin inflammation and the development of hematopoietic cells. Little is known about its other biological functions and clinical implications. These cytokines display pro-inflammatory activities in different tissues, including skin.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NYY1
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 50604
Molecular Weight (g/mol) 35.2 kDa
Name Human IL-20
Quantity 500 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias cytokine Zcyto10; four alpha helix cytokine; four alpha helix cytokine ZCYTO10; four helix bundle cytokine 10; il 20; IL10D; Il20; IL-20; ILN; Interleukin; interleukin 20; interleukin-20; MGC96907; PRO1801; UNQ852; UNQ852/PRO1801; ZCYTO10
Common Name IL-20
Gene Symbol Il20
Biological Activity Determined by its ability to stimulate the proliferation of HEK-Blue IL-19 TM reporter cells.
Conjugate Unconjugated
Recombinant Recombinant
Sequence MLKTLNLGSC VIATNLQEIR NGFSEIRGSV QAKDGNIDIR ILRRTESLQD TKPANRCCLL RHLLRLYLDR VFKNYQTPDH YTLRKISSLA NSFLTIKKDL RLCHAHMTCH CGEEAMKKYS QILSHFEKLE PQAAVVKALG ELDILLQWME ETE
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.