missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 14 publications
2094.75 NOK - 19856.55 NOK
Specifications
| Antigen | IL-2 |
|---|---|
| Clone | 5334 |
| Concentration | LYOPH |
| Dilution | Neutralization 0.015-0.03 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready |
| Applications | Neutralization, Flow Cytometry, Immunocytochemistry, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18763317
![]() |
R&D Systems
MAB202-SP |
25 μg |
2215.00 NOK 2094.75 NOK / 25µg Save 120.25 NOK 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18774368
![]() |
R&D Systems
MAB202-100 |
100 μg |
5145.00 NOK 4866.75 NOK / 100µg Save 278.25 NOK 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18712698
![]() |
R&D Systems
MAB202-500 |
500 μg |
20995.00 NOK 19856.55 NOK / 500µg Save 1138.45 NOK 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-2 Monoclonal specifically detects IL-2 in Human samples. It is validated for Neutralization, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
| IL-2 | |
| LYOPH | |
| Neutralization, Flow Cytometry, Immunocytochemistry, CyTOF | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| aldesleukin, IL2, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, Proleukin, T cell growth factor, T-cell growth factor, TCGF | |
| IL2 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 5334 | |
| Neutralization 0.015-0.03 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Human | |
| NP_000577 | |
| 3558 | |
| E. coli-derived recombinant human IL-2 Ala21-Thr153 Accession # NP_000577 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IL-2 in direct ELISAs. Does not cross-react with recombinant IL-2 from mouse, rat, pig, or cotton rat. |
Spot an opportunity for improvement?
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.