missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-13 R alpha 2 Antibody, R&D Systems™
Rabbit Monoclonal Antibody
2145.00 NOK - 4840.00 NOK
Specifications
| Antigen | IL-13R alpha 2 |
|---|---|
| Clone | 2725C |
| Dilution | Western Blot 0.5 ug/mL, Simple Western 0.5 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18159010
|
R&D Systems
MAB6142-100 |
100 μg |
4840.00 NOK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18192511
|
R&D Systems
MAB6142-SP |
25 μg |
2145.00 NOK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-13R alpha 2 Monoclonal antibody specifically detects IL-13R alpha 2 in Human samples. It is validated for Western Blot, Western BlotSpecifications
| IL-13R alpha 2 | |
| Western Blot 0.5 ug/mL, Simple Western 0.5 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| cancer/testis antigen 19, CD213a2, CD213a2 antigen, CT19, IL-13 receptor subunit alpha-2, IL13BP, IL13R, IL-13R, IL13R alpha 2, IL-13R subunit alpha-2, IL13RA2, IL-13RA2, IL-13R-alpha-2, interleukin 13 binding protein, interleukin 13 receptor alpha 2 chain, interleukin 13 receptor, alpha 2, interleukin-13 receptor subunit alpha-2, Interleukin-13-binding protein | |
| Chinese Hamster Ovary cell line CHO-derived Human IL-13 R alpha 2, Met1-Leu342, Accession # NP_000631 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 2725C | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| NP_000631 | |
| 3598 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title