missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-12/IL-23 p40 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB609-SP
1479.00 NOK valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
IL-12/IL-23 p40 Monoclonal specifically detects IL-12/IL-23 p40 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
| IL-12/IL-23 p40 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL12 p40, IL-12 subunit p40, IL12B, IL-12B, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2, NKSF2IL12, subunit p40 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 3593 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| ELISA | |
| 31052 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P29460 | |
| IL12B | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-12/IL-23 p40 free monomer Ile23-Ser328 Accession # P29460 | |
| 25 μg | |
| Primary | |
| Detects human IL-12/IL-23 p40 in ELISAs. In ELISAs, this antibody shows less than 5% cross-reactivity with recombinant human (rh) IL-12 (p35/p40 heterodimer) and rhIL-23 (p19/p40 heterodimer). No significant cross-reactivity or interference was observed with rhIL-1 alpha , rmIL-1 alpha , rhIL-1 beta , rmIL-1 beta , rhIL-1ra, rhIL-1 RI, rhIL-1 RII, rhIL-2, rhIL-2 R alpha , rhIL-3, rmIL-3, rhIL-4, rmIL-4, rhIL-4 R, rhIL-5, rmIL-5, rhIL-5 R beta , rhIL-6, rhIL-6 R, rmIL-6, rhIL-7, rmIL-7, rhIL-8, rhIL-9, rmIL-9, rhIL-10, rmIL-10, rhIL-11, rmIL-12, rmIL-12 p40, rhIL-13, or rmIL-13. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction