missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-1 RI APC-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
2948.40 NOK - 5389.65 NOK
Specifications
| Antigen | IL-1 RI |
|---|---|
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | APC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18787157
![]() |
R&D Systems
FAB269A-025 |
25 Tests |
3120.00 NOK 2948.40 NOK / 25 tests Save 171.60 NOK 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18718817
![]() |
R&D Systems
FAB269A-100 |
100 Tests |
5700.00 NOK 5389.65 NOK / 100 tests Save 310.35 NOK 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-1 RI Polyclonal specifically detects IL-1 RI in Human samples. It is validated for Flow Cytometry.Specifications
| IL-1 RI | |
| Flow Cytometry | |
| APC | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| 3554 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-1 RI | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| Flow Cytometry 10 uL/10^6 cells | |
| Polyclonal | |
| Goat | |
| Human | |
| CD121 antigen-like family member A, CD121a, CD121a antigen, IL1R, IL1R1, IL1RI, IL-1RI, IL1RT1, IL-1RT1, IL-1RT-1, Interleukin 1 Receptor 1, interleukin 1 receptor, type I, interleukin receptor 1, interleukin-1 receptor type 1, p80 | |
| IL1R1 | |
| IgG | |
| Affinity Purified | |
| Detects human IL-1 RI in direct ELISAs and Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title