missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human IHPK2 Full-length ORF (AAH65533.1, 1 a.a. - 426 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00051447-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16190033

  • 6245.00 NOK / 25µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a protein that belongs to the inositol phosphokinase (IPK) family. This protein is likely responsible for the conversion of inositol hexakisphosphate (InsP6) to diphosphoinositol pentakisphosphate (InsP7/PP-InsP5). It may also convert 1,3,4,5,6-pentakisphosphate (InsP5) to PP-InsP4 and affect the growth suppressive and apoptotic activities of interferon-beta in some ovarian cancers. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]

Sequence: MSPAFRAMDVEPRAKGVLLEPFVHQVGGHSCVLRFNETTLCKPLVPREHQFYETLPAEMRKFTPQYKGVVSVRFEEDEDRNLCLIAYPLKGDHGIVDIVDNSDCEPKSKLLRWTTNKKHHVLETEKTPKDWVRQHRKEEKMKSHKLEEEFEWLKKSEVLYYTVEKKWNISSQLKHYNPWSMKCHQQQLQRMKENAKHRNQYKFILLENLTSRYEVPCVLDLKMGTRQHGDDASEEKAANQIRKCQQSTSAVIGVRVCGMQVYQAGSGQLMFMNKYHGRKLSVQGFKEALFQFFHNGRYLRRELLGPVLKKLTELKAVLERQESYRFYSSSLLVIYDGKERPEVVLDSDAEDLEDLSEESADESAGAYAYKPIGASSVDVRMIDFAHTTCRLYGEDTVVHEGQDAGYIFGLQSLIDIVTEISEESGE
Specifications

Specifications

AAH65533.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
75.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
IHPK2/PiUS
IP6K2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
51447
IHPK2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSPAFRAMDVEPRAKGVLLEPFVHQVGGHSCVLRFNETTLCKPLVPREHQFYETLPAEMRKFTPQYKGVVSVRFEEDEDRNLCLIAYPLKGDHGIVDIVDNSDCEPKSKLLRWTTNKKHHVLETEKTPKDWVRQHRKEEKMKSHKLEEEFEWLKKSEVLYYTVEKKWNISSQLKHYNPWSMKCHQQQLQRMKENAKHRNQYKFILLENLTSRYEVPCVLDLKMGTRQHGDDASEEKAANQIRKCQQSTSAVIGVRVCGMQVYQAGSGQLMFMNKYHGRKLSVQGFKEALFQFFHNGRYLRRELLGPVLKKLTELKAVLERQESYRFYSSSLLVIYDGKERPEVVLDSDAEDLEDLSEESADESAGAYAYKPIGASSVDVRMIDFAHTTCRLYGEDTVVHEGQDAGYIFGLQSLIDIVTEISEESGE
RUO
IP6K2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers