missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human IGSF1 Full-length ORF (NP_991402.1, 1 a.a. - 242 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003547-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16196711

  • 4115.00 NOK / 10µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Members of the immunoglobulin (Ig) superfamily (see MIM 147100), which includes IGSF1, have a variety of functions, but all appear to play a role in cell recognition and the regulation of cell behavior.[supplied by OMIM]

Sequence: MTLDRPGEGATMLKTFTVLLFCIRMSLGMTSIVMDPQPELWIESNYPQAPWENITLWCRSPSRISSKFLLLKDKTQMTWIRPSHKTFQVSFLIGALTESNAGLYRCCYWKETGWSKPSKVLELEAPGQLPKPIFWIQAETPALPGCNVNILCHGWLQDLVFMLFKEGYAEPVDYQVPTGTMAIFSIDNLTPEDEGVYICRTHIQMLPTLWSEPSNPLKLVVAGGCGYGCWHLAIVVPGIMAG
Specifications

Specifications

NP_991402.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
53.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
IGCD1/IGDC1/INHBP/KIAA0364/MGC75490/PGSF2
IGSF1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
3547
IGSF1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTLDRPGEGATMLKTFTVLLFCIRMSLGMTSIVMDPQPELWIESNYPQAPWENITLWCRSPSRISSKFLLLKDKTQMTWIRPSHKTFQVSFLIGALTESNAGLYRCCYWKETGWSKPSKVLELEAPGQLPKPIFWIQAETPALPGCNVNILCHGWLQDLVFMLFKEGYAEPVDYQVPTGTMAIFSIDNLTPEDEGVYICRTHIQMLPTLWSEPSNPLKLVVAGGCGYGCWHLAIVVPGIMAG
RUO
IGSF1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers