missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human IGLC2 Full-length ORF (AAH93098.1, 1 a.a. - 235 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003538-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16136711

  • 4115.00 NOK / 10µg
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MASFPLLLTLLTHCAGSWAQSVLTQPPSASGTPGQRVPISCSGSSSNIGSNTVNWYQQFPGTAPKLLIYSNNQRPSGVPDRFSGSKSGTSASLAISGLQSEDDAVYHCATWDDNLNSWVFGGGTKLTVLSQPKAAPSVTLFPPSSEELQANKATLVCLISDFYPGAVTVAWKADSSPVKAGVETTTPSKQSNNKYAASSYLSLTPEQWKSHKSYSCQVTHEGSTVEKTVAPTECS
Specifications

Specifications

AAH93098.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
51.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
IGLC/MGC20392/MGC45681
IGLC2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
3538
IGLC2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MASFPLLLTLLTHCAGSWAQSVLTQPPSASGTPGQRVPISCSGSSSNIGSNTVNWYQQFPGTAPKLLIYSNNQRPSGVPDRFSGSKSGTSASLAISGLQSEDDAVYHCATWDDNLNSWVFGGGTKLTVLSQPKAAPSVTLFPPSSEELQANKATLVCLISDFYPGAVTVAWKADSSPVKAGVETTTPSKQSNNKYAASSYLSLTPEQWKSHKSYSCQVTHEGSTVEKTVAPTECS
RUO
IGLC2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers