missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human IGKV1OR2-108 Partial ORF (CAA36171.1, 1 a.a. - 95 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00028862-Q01.25ug
Dettagli aggiuntivi : Weight : 0.00010kg
Descrizione
Sequence: DIQVTQSPSSLSASVGDRVTITCRASQGISNGLSWYQQKPGQAPTLLIYAASSLQSGVPSRFSGSGSGTDFTLTISSLQPEDVATYYCLQDYTTPSpecifica
CAA36171.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.19kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DIQVTQSPSSLSASVGDRVTITCRASQGISNGLSWYQQKPGQAPTLLIYAASSLQSGVPSRFSGSGSGTDFTLTISSLQPEDVATYYCLQDYTTP | |
RUO | |
IGKV1OR2-108 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
28862 | |
IGKV1OR2-108 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IGKV1OR2108/IGO1 | |
IGKV1OR2-108 | |
Recombinant | |
wheat germ expression system |