missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human IGF2R Partial ORF (NP_000867, 121 a.a. - 220 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003482-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16126661

  • 6245.00 NOK / 25µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a receptor for both, insulin-like growth factor 2 (IGF2) and mannose 6-phosphate (M6P). The IGF2 and M6P binding sites are located on different segments of the receptor. This receptor functions in the intracellular trafficking of lysosomal enzymes, the activation of transforming growth factor beta, and the degradation of IGF2. In mice, the homologous gene is expressed only from the maternal chromosome. [provided by RefSeq]

Sequence: GTNHRVQSSIAFLCGKTLGTPEFVTATECVHYFEWRTTAACKKDIFKANKEVPCYVFDEELRKHDLNPLIKLSGAYLVDDSDPDTSLFINVCRDIDTLRD
Specifications

Specifications

NP_000867
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GTNHRVQSSIAFLCGKTLGTPEFVTATECVHYFEWRTTAACKKDIFKANKEVPCYVFDEELRKHDLNPLIKLSGAYLVDDSDPDTSLFINVCRDIDTLRD
RUO
IGF2R
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
3482
IGF2R (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD222/CIMPR/M6P-R/MPR1/MPRI
IGF2R
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers