missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IGF-II R/IGF2R Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB24471
This item is not returnable.
View return policy
Description
IGF-II R/IGF2R Monoclonal specifically detects IGF-II R/IGF2R in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
| IGF-II R/Mannose 6 Phosphate Receptor (Cation independent) | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 300 kDa mannose 6-phosphate receptor, cation-independent mannose-6 phosphate receptor, cation-independent mannose-6-phosphate receptor, CD222, CD222 antigen, CI Man-6-P receptor, CIMPR, CI-MPR, IGF2R, IGF-II receptor, IGFIIR, IGF-IIR, insulin-like growth factor 2 receptorM6P/IGF2R, Insulin-like growth factor II receptor, M6P/IGF2 receptor, M6PR, M6P-R, MPR 300, MPR1, MPRI, MPRIM6PR | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3482 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| ELISA | |
| 372616 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P11717 | |
| IGF2R | |
| Mouse myeloma cell line NS0-derived recombinant human IGF-II R Ser1510-Phe2108 Accession # P11717 | |
| 500 μg | |
| Primary | |
| Detects human IGF-II R in ELISAs. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction