missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human IFNGR2 Partial ORF (NP_005525.2, 28 a.a. - 127 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003460-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16189894

  • 6245.00 NOK / 25µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene (IFNGR2) encodes the non-ligand-binding beta chain of the gamma interferon receptor. Human interferon-gamma receptor is a heterodimer of IFNGR1 and IFNGR2. Defects in IFNGR2 are a cause of mendelian susceptibility to mycobacterial disease (MSMD), also known as familial disseminated atypical mycobacterial infection. MSMD is a genetically heterogeneous disease with autosomal recessive, autosomal dominant or X-linked inheritance. [provided by RefSeq]

Sequence: SQLPAPQHPKIRLYNAEQVLSWEPVALSNSTRPVVYQVQFKYTDSKWFTADIMSIGVNCTQITATECDFTAASPSAGFPMDFNVTLRLRAELGALHSAWV
Specifications

Specifications

NP_005525.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SQLPAPQHPKIRLYNAEQVLSWEPVALSNSTRPVVYQVQFKYTDSKWFTADIMSIGVNCTQITATECDFTAASPSAGFPMDFNVTLRLRAELGALHSAWV
RUO
IFNGR2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
3460
IFNGR2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AF-1/IFGR2/IFNGT1
IFNGR2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers