Learn More
Human IFN-beta Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB814R-100
 View more versions of this product
Product Code. 18712549
  • 4610.00 NOK / 100µg
Description
IFN-beta Monoclonal specifically detects IFN-beta in Human samples. It is validated for Neutralization.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IFN-beta
Monoclonal
Unconjugated
Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956
Mouse
Protein A or G purified from cell culture supernatant
RUO
3456
Human
Purified
Neutralization
76703R
Neutralization 7-21 ug/mL
P01574
IFNB1
E. coli-derived recombinant human IFN-beta Met22-Asn187 Accession # P01574
100 μg
Primary
Detects human IFN-beta in direct ELISAs.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1