Learn More
Human IFN-beta Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB814R-500
This item is not returnable.
View return policy
Description
IFN-beta Monoclonal specifically detects IFN-beta in Human samples. It is validated for Neutralization.
Specifications
| IFN-beta | |
| 76703R | |
| Neutralization 7-21 ug/mL | |
| P01574 | |
| IFNB1 | |
| E. coli-derived recombinant human IFN-beta Met22-Asn187 Accession No. P01574 | |
| 500 ug | |
| Primary | |
| Detects human IFN-beta in direct ELISAs. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Monoclonal | |
| Unconjugated | |
| Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 um filtered solution in PBS. with No Preservative | |
| Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3456 | |
| Human | |
| Purified |
Product Content Correction
Product Title
For Research Use Only