Learn More
Abnova™ Human ICT1 Partial ORF (NP_001536, 107 a.a. - 206 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00003396-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The adult colon epithelium contains 3 differentiated cell types that arise from a multipotent stem cell. Deviation from the normal maturation pathway by neoplastic transformation is thought to initiate in stem cells or their early descendants. One potential marker is ICT1 whose mRNA and protein were more highly expressed in undifferentiated than in differentiated cells. [provided by RefSeq]
Sequence: TAEWIAEPVRQKIAITHKNKINRLGELILTSESSRYQFRNLADCLQKIRDMITEASQTPKEPTKEDVKLHRIRIENMNRERLRQKRIHSAVKTSRRVDMDSpecifications
NP_001536 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TAEWIAEPVRQKIAITHKNKINRLGELILTSESSRYQFRNLADCLQKIRDMITEASQTPKEPTKEDVKLHRIRIENMNRERLRQKRIHSAVKTSRRVDMD | |
RUO | |
ICT1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3396 | |
ICT1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DS-1 | |
ICT1 | |
Recombinant | |
wheat germ expression system |