missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ICAM-1 (CD54) Fc Chimera, Recombinant Protein
HEK293 cells Recombinant Protein
Brand: Invitrogen™ A42523
Additional Details : Weight : 0.00500kg
Description
ICAM-1 (CD54) is an 85-110 kDa single-chain type 1 integral membrane glycoprotein with an extracellular domain of five immunoglobulin superfamily repeats, a transmembrane region and a cytoplasmic domain. ICAM-1 has 7 potential N-linked glycosylation sites and shares considerable amino acid sequence homology with ICAM-3 (CD50) and ICAM-2 (CD102). ICAM-1 binds to integrins of type CD11a/CD18 (leukocyte adhesion molecule, LFA-1), or CD11b/CD18 (Mac-1) and is exploited by Rhinovirus as a receptor. ICAM-1 is expressed by activated endothelial cells and detected on epithelial cells, fibroblasts, chondrocytes, B lymphocytes, T lymphocytes (low), monocytes, macrophages, dendritic cells and neutrophils, with lower levels that increase upon inflammation. ICAM-1 is also detected in some carcinoma and melanoma cells. Soluble ICAM-1 is detectable in the plasma and is elevated in patients with various inflammatory syndromes.Specifications
P05362 | |
protein with 0.02% Tween 80, 140 mM NaCl, 2.7 mM KCl, 5% mannitol, 5% trehalose and no preservative | |
Human ICAM-1 (CD54) Fc Chimera | |
HEK293 cells | |
RUO | |
BB2; CD54; cell surface glycoprotein P3.58; ICAM; Icam1; ICAM-1; ICAM-1; CD54 homolog; Intercellular adhesion molecule 1; intercellular adhesion molecule 1 (CD54), human rhinovirus receptor; intercellular adhesion molecule-1; intercellular adhesion molecule-1 precursor; intercellular adhesion molecule-like; LOC100345640; Ly-47; Major group rhinovirus receptor; MALA-2; MyD10; P3.58; sCD54; soluble CD54 | |
Human | |
Recombinant | |
≥ 90% |
Control, ELISA, Functional Assay, Immunohistochemistry, Western Blot | |
3383 | |
50 μg | |
-20°C | |
<1 EU/μg | |
ICAM1 | |
Unconjugated | |
Lyophilized |