missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ICAM-1/CD54 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 9 publications
Brand: R&D Systems BBA17
This item is not returnable.
View return policy
Description
ICAM-1/CD54 Polyclonal specifically detects ICAM-1/CD54 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Knockout Validated.
Specifications
| ICAM-1/CD54 | |
| Polyclonal | |
| Unconjugated | |
| BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM1, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58 | |
| Goat | |
| >50% SDS-PAGE | |
| RUO | |
| 3383 | |
| Reconstitute in 0.5 mL of sterile water. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoassay | |
| LYOPH | |
| Western Blot 1:1000 dilution, Immunohistochemistry 1:300 dilution, Immunocytochemistry 1-25 ug/mL, Knockout Validated 1:1000 ug/mL | |
| ICAM1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human ICAM-1/CD54 Extracellular domain | |
| 500 μL | |
| Primary | |
| Detects human ICAM-1/CD54 in direct ELISAs and Western blots. ICAM-1/CD54 has been screened using CHO cells transfected with cDNAs for ICAM-1, VCAM-1, and E-Selectin. ICAM-1/CD54 was shown to be only reactive with ICAM-1. | |
| Human | |
| Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction