missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HSPA6 Partial ORF (NP_002146.2, 544 a.a. - 643 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003310-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16189684

  • 4115.00 NOK / 10µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: LEAHVFHVKGSLQEESLRDKIPEEDRRKMQDKCREVLAWLEHNQLAEKEEYEHQKRELEQICRPIFSRLYGGPGVPGGSSCGTQARQGDPSTGPIIEEVD
Specifications

Specifications

NP_002146.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LEAHVFHVKGSLQEESLRDKIPEEDRRKMQDKCREVLAWLEHNQLAEKEEYEHQKRELEQICRPIFSRLYGGPGVPGGSSCGTQARQGDPSTGPIIEEVD
RUO
HSPA6
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
3310
HSPA6 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HSPA6
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers