Learn More
Abnova™ Human HSF2BP Full-length ORF (NP_008962.1, 1 a.a. - 334 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00011077-P01.25ug
Additional Details : Weight : 0.00010kg
Description
HSF2 binding protein (HSF2BP) associates with HSF2. The interaction occurs between the trimerization domain of HSF2 and the amino terminal hydrophilic region of HSF2BP that comprises two leucine zipper motifs. HSF2BP may therefore be involved in modulating HSF2 activation. [provided by RefSeq]
Sequence: MGEAGAAEEACRHMGTKEEFVKVRKKDLERLTTEVMQIRDFLPRILNGEVLESFQKLKIVEKNLERKEQELEQLKMDCEHFKARLETVQADNIREKKEKLALRQQLNEAKQQLLQQAEYCTEMGAAACTLLWGVSSSEEVVKAILGGDKALKFFSITGQTMESFVKSLDGDVQELDSDESQFVFALAGIVTNVAAIACGREFLVNSSRVLLDTILQLLGDLKPGQCTKLKVLMLMSLYNVSINLKGLKYISESPGFIPLLWWLLSDPDAEVCLHVLRLVQSVVLEPEVFSKSASEFRSSLPLQRILAMSKSRNPRLQTAAQELLEDLRTLEHNVSpecifications
NP_008962.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
64kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HSF2BP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
11077 | |
HSF2BP (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGEAGAAEEACRHMGTKEEFVKVRKKDLERLTTEVMQIRDFLPRILNGEVLESFQKLKIVEKNLERKEQELEQLKMDCEHFKARLETVQADNIREKKEKLALRQQLNEAKQQLLQQAEYCTEMGAAACTLLWGVSSSEEVVKAILGGDKALKFFSITGQTMESFVKSLDGDVQELDSDESQFVFALAGIVTNVAAIACGREFLVNSSRVLLDTILQLLGDLKPGQCTKLKVLMLMSLYNVSINLKGLKYISESPGFIPLLWWLLSDPDAEVCLHVLRLVQSVVLEPEVFSKSASEFRSSLPLQRILAMSKSRNPRLQTAAQELLEDLRTLEHNV | |
RUO | |
HSF2BP | |
Recombinant | |
wheat germ expression system |