missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HSD17B7 Full-length ORF (NP_057455.1, 1 a.a. - 341 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00051478-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16140063

  • 4115.00 NOK / 10µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The 17-beta-hydroxysteroid dehydrogenase enzyme (EC 1.1.1.62) oxidizes or reduces estrogens and androgens in mammals and regulates the biologic potency of these steroids.[supplied by OMIM]

Sequence: MRKVVLITGASSGIGLALCKRLLAEDDELHLCLACRNMSKAEAVCAALLASHPTAEVTIVQVDVSNLQSVFRASKELKQRFQRLDCIYLNAGIMPNPQLNIKALFFGLFSRKVIHMFSTAEGLLTQGDKITADGLQEVFETNVFGHFILIRELEPLLCHSDNPSQLIWTSSRSARKSNFSLEDFQHSKGKEPYSSSKYATDLLSVALNRNFNQQGLYSNVACPGTALTNLTYGILPPFIWTLLMPAILLLRFFANAFTLTPYNGTEALVWLFHQKPESLNPLIKYLSATTGFGRNYIMTQKMDLDEDTAEKFYQKLLELEKHIRVTIQKTDNQARLSGSCL
Specifications

Specifications

NP_057455.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
64.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC12523/MGC75018/PRAP/SDR37C1
HSD17B7
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
51478
HSD17B7 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MRKVVLITGASSGIGLALCKRLLAEDDELHLCLACRNMSKAEAVCAALLASHPTAEVTIVQVDVSNLQSVFRASKELKQRFQRLDCIYLNAGIMPNPQLNIKALFFGLFSRKVIHMFSTAEGLLTQGDKITADGLQEVFETNVFGHFILIRELEPLLCHSDNPSQLIWTSSRSARKSNFSLEDFQHSKGKEPYSSSKYATDLLSVALNRNFNQQGLYSNVACPGTALTNLTYGILPPFIWTLLMPAILLLRFFANAFTLTPYNGTEALVWLFHQKPESLNPLIKYLSATTGFGRNYIMTQKMDLDEDTAEKFYQKLLELEKHIRVTIQKTDNQARLSGSCL
RUO
HSD17B7
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers