Learn More
Abnova™ Human HS3ST4 Partial ORF (NP_006031.1, 358 a.a. - 455 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009951-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes the enzyme heparan sulfate D-glucosaminyl 3-O-sulfotransferase 4. This enzyme generates 3-O-sulfated glucosaminyl residues in heparan sulfate. Cell surface heparan sulfate is used as a receptor by herpes simplex virus type 1 (HSV-1), and expression of this gene is thought to play a role in HSV-1 pathogenesis. [provided by RefSeq]
Sequence: ERLIVDPAGEMAKVQDFLGLKRVVTKKHFYFNKTKGFPCLKKPEDSSAPRCLGKSKGRTHPRIDPDVIHRLRKFYKPFNLMFYQMTGQDFQWEQEEGDSpecifications
NP_006031.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ERLIVDPAGEMAKVQDFLGLKRVVTKKHFYFNKTKGFPCLKKPEDSSAPRCLGKSKGRTHPRIDPDVIHRLRKFYKPFNLMFYQMTGQDFQWEQEEGD | |
RUO | |
HS3ST4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9951 | |
HS3ST4 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
3-OST-4/30ST4/3OST4 | |
HS3ST4 | |
Recombinant | |
wheat germ expression system |